accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51449-86-6

H3CNNNN

CAS RN 51449-86-6

IUPAC Name: 5-(2-methylphenyl)-1H-1,2,3,4-tetrazol-1-ide
Synonyms: 5-(2-methylphenyl)-1H-1,2,3,4-tetrazol-1-ide
Molecular Weight (g/mol): 159.17
Molecular Formula: C8H7N4
InChi Key: WXCHXJGVYSYFLZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=NN=N[N-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results