accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5145-99-3

IUPAC Name: 2-(ethylsulfanyl)propane
Synonyms: ethyl isopropyl sulfide
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: NZUQQADVSXWVNW-UHFFFAOYSA-N
SMILES: CCSC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results