accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51450-36-3

IUPAC Name: 2,2-dimethyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Synonyms: 2,2-dimethyl-4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-ol
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: GAQDIYMHBQNXLE-UHFFFAOYNA-N
SMILES: CC1(C)OCC2OC=CC(O)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results