accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5146-66-7

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dienenitrile
Synonyms: (2E)-3,7-dimethylocta-2,6-dienenitrile
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: HLCSDJLATUNSSI-JXMROGBWSA-N
SMILES: CC(C)=CCC\C(C)=C\C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results