accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OON

CAS RN 51473-74-6

IUPAC Name: 7-(diethylamino)-2-oxo-2H-chromene-3-carbonitrile
Synonyms: 7-(diethylamino)-2-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 242.28
Molecular Formula: C14H14N2O2
InChi Key: LOUYEVRVQGFIFB-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C#N)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results