accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClOOONH2H3CCH3H3C(S)CH3

CAS RN 51482-39-4

IUPAC Name: methyl (2S)-2-amino-3-[4-(tert-butoxy)phenyl]propanoate hydrochloride
Synonyms: methyl (2S)-2-amino-3-[4-(tert-butoxy)phenyl]propanoate hydrochloride
Molecular Weight (g/mol): 287.78
Molecular Formula: C14H22ClNO3
InChi Key: PAFVAMWJVIIMQK-YDALLXLXSA-N
SMILES: Cl.COC(=O)[C@@H](N)CC1=CC=C(OC(C)(C)C)C=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)