accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51484-06-1

IUPAC Name: 6-bromo-4-oxo-4H-chromene-2-carboxylic acid
Synonyms: 6-bromo-4-oxochromene-2-carboxylic acid
Molecular Weight (g/mol): 269.05
Molecular Formula: C10H5BrO4
InChi Key: QSBZDBNPXSVVHH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=CC(Br)=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results