accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHOO

CAS RN 515-30-0

IUPAC Name: 2-hydroxy-2-phenylpropanoic acid
Synonyms: (+-)-atrolactic acid (1)-2-phenyllactic acid hydroxymethylmandelic acid atrolactic acid, (+-)- atrolactinic acid dl-atrolactic acid (+-)-isomer atrolactic acid dl-atrolactate (S)-atrolactic acid atrolactinate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: NWCHELUCVWSRRS-UHFFFAOYNA-N
SMILES: CC(O)(C(O)=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results