accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClClCl

CAS RN 515-84-4

IUPAC Name: ethyl 2,2,2-trichloroacetate
Synonyms: ethyl trichloroacetate
Molecular Weight (g/mol): 191.43
Molecular Formula: C4H5Cl3O2
InChi Key: SJMLNDPIJZBEKY-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Cl)(Cl)Cl
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)