accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 515-96-8

NH2HNONH2O

CAS RN 515-96-8

IUPAC Name: 1-(hydrazinecarbonyl)formamide
Synonyms: 1-(hydrazinecarbonyl)formamide
Molecular Weight (g/mol): 103.08
Molecular Formula: C2H5N3O2
InChi Key: MOKRDWKSHLLYKM-UHFFFAOYSA-N
SMILES: NNC(=O)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results