accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51513-29-2

IUPAC Name: methyl 2-carbamoylacetate
Synonyms: methyl 2-carbamoylacetate
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: LSNSJCKGQREPDW-UHFFFAOYSA-N
SMILES: COC(=O)CC(N)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)