accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHONH

CAS RN 515140-26-8

IUPAC Name: [4-(cyclopropylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(cyclopropylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 205.02
Molecular Formula: C10H12BNO3
InChi Key: WCRPDYXXIVYAAJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)NC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results