accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51516-67-7

IUPAC Name: 5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Synonyms: 5-amino-1-(4-chlorophenyl)pyrazole-4-carbonitrile
Molecular Weight (g/mol): 218.64
Molecular Formula: C10H7ClN4
InChi Key: WIWSALMJHPGFDY-UHFFFAOYSA-N
SMILES: NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results