accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51537-21-4

CH3H3CCH3OOOCH3H3CCH3H2NHCl

CAS RN 51537-21-4

IUPAC Name: tert-butyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Synonyms: tert-butyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Molecular Weight (g/mol): 253.77
Molecular Formula: C11H24ClNO3
InChi Key: RDWZQVGVBTYCBD-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OCC(N)C(=O)OC(C)(C)C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)