accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5158-46-3

IUPAC Name: chlorozincylium methanide
Synonyms: chlorozincylium methanide
Molecular Weight (g/mol): 115.87
Molecular Formula: CH3ClZn
InChi Key: NMLXKNNXODLJIN-UHFFFAOYSA-M
SMILES: [CH3-].Cl[Zn+]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results