accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 516-03-0

FeOOOO

CAS RN 516-03-0

IUPAC Name: λ²-iron(2+) oxalate
Synonyms: ferrous oxalate
Molecular Weight (g/mol): 143.86
Molecular Formula: C2FeO4
InChi Key: OWZIYWAUNZMLRT-UHFFFAOYSA-L
SMILES: [Fe++].[O-]C(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results