accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOOHO

CAS RN 516-05-2

IUPAC Name: 2-methylpropanedioic acid
Synonyms: methylmalonic acid
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: ZIYVHBGGAOATLY-UHFFFAOYSA-N
SMILES: CC(C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results