accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHONH2CH3

CAS RN 516-06-3

IUPAC Name: 2-amino-3-methylbutanoic acid
Synonyms: valin|valina|Hval|DL-valine
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: KZSNJWFQEVHDMF-UHFFFAOYSA-N
SMILES: CC(C)C(N)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Grade 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)