CAS RN 51605-32-4
CAS RN 51605-32-4
IUPAC Name:
ethyl 5-methyl-1H-imidazole-4-carboxylate
Synonyms:
ethyl 5-methyl-1H-imidazole-4-carboxylate
Molecular Weight (g/mol):
154.17
Molecular Formula:
C7H10N2O2
InChi Key:
VLDUBDZWWNLZCU-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)NC=N1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ethyl 4-methyl-5-imidazolecarboxylate, 98%
CAS: 51605-32-4 Molecular Formula: C7H10N2O2 Molecular Weight (g/mol): 154.17 MDL Number: MFCD00005199 InChI Key: VLDUBDZWWNLZCU-UHFFFAOYSA-N Synonym: ethyl 4-methyl-5-imidazolecarboxylate,ethyl 4-methyl-1h-imidazole-5-carboxylate,ethyl-4-methyl-5-imidazolecarboxylate,1h-imidazole-5-carboxylic acid, 4-methyl-, ethyl ester,1h-imidazole-4-carboxylic acid, 5-methyl-, ethyl ester,4-carboethoxy-5-methylimidazole,ethyl 4-methylimidazole-5-carboxylate,ethyl 5-methyl-4-imidazolecarboxylate,5-methyl-3h-imidazole-4-carboxylic acid ethyl ester,ethyl 5-methylimidazole-4-carboxylate PubChem CID: 99200 IUPAC Name: ethyl 5-methyl-1H-imidazole-4-carboxylate SMILES: CCOC(=O)C1=C(C)NC=N1