accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51627-47-5

NH2HNNO

CAS RN 51627-47-5

IUPAC Name: 3-amino-2-(morpholin-4-yl)pyridin-1-ium
Synonyms: 3-amino-2-(morpholin-4-yl)pyridin-1-ium
Molecular Weight (g/mol): 180.23
Molecular Formula: C9H14N3O
InChi Key: DHYQHRDXKCKJAN-UHFFFAOYSA-O
SMILES: NC1=CC=C[NH+]=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results