accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COONOCl

CAS RN 51630-58-1

IUPAC Name: cyano(3-phenoxyphenyl)methyl 2-(4-chlorophenyl)-3-methylbutanoate
Synonyms: tirade pydrin, (R-(R*,S*))- aqmatrine epa shaughnessy code: 109301 pydrin, (S-(R*,S*))- (s-(r*,s*))-pydrin sumibac (S-(R*,S*))-isomer pydrin evercide 2362 sumifly
Molecular Weight (g/mol): 419.91
Molecular Formula: C25H22ClNO3
InChi Key: NYPJDWWKZLNGGM-UHFFFAOYNA-N
SMILES: CC(C)C(C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results