accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51632-28-1

IUPAC Name: 2-(4-ethylphenyl)acetonitrile
Synonyms: 2-(4-ethylphenyl)acetonitrile
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: NCPKDFDQDZMXCO-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CC#N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results