accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOOHOHO

CAS RN 51649-83-3

IUPAC Name: 6-amino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 6-amino-3',6'-dihydroxyspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 347.33
Molecular Formula: C20H13NO5
InChi Key: YOAWSYSKQHLFPM-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)OC3(C2=C1)C1=CC=C(O)C=C1OC1=CC(O)=CC=C31
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results