accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5166-67-6

IUPAC Name: ethyl 1-methylpiperidine-3-carboxylate
Synonyms: ethyl 1-methylpiperidine-3-carboxylate
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: VFJJNMLPRDRTCO-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN(C)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results