accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3I

CAS RN 51660-65-2

IUPAC Name: 4-iodo-1,3,5-trimethyl-1H-pyrazole
Synonyms: 4-iodo-1,3,5-trimethylpyrazole
Molecular Weight (g/mol): 236.06
Molecular Formula: C6H9IN2
InChi Key: RUOGLHMRBURIEQ-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(I)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results