accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOO(S)OHN

CAS RN 51693-17-5

IUPAC Name: [(2S)-5-oxopyrrolidin-2-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: [(2S)-5-oxopyrrolidin-2-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 269.32
Molecular Formula: C12H15NO4S
InChi Key: AMZNHHZJURKRFX-JTQLQIEISA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CCC(=O)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results