accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5170-68-3

IUPAC Name: 2,1,3-benzothiadiazole-4-carbaldehyde
Synonyms: 2,1,3-benzothiadiazole-4-carbaldehyde
Molecular Weight (g/mol): 164.18
Molecular Formula: C7H4N2OS
InChi Key: ANVJARPTPIVPRC-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC2=NSN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results