accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBrSBr

CAS RN 51751-44-1

IUPAC Name: 3,3'-dibromo-2,2'-bithiophene
Synonyms: 3,3'-dibromo-2,2'-bithiophene
Molecular Weight (g/mol): 324.05
Molecular Formula: C8H4Br2S2
InChi Key: KBRZCEVRNLKHAZ-UHFFFAOYSA-N
SMILES: BrC1=C(SC=C1)C1=C(Br)C=CS1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)