accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51760-21-5

IUPAC Name: 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 273.08
Molecular Formula: C10H9BrO4
InChi Key: QUJINGKSNJNXEB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(Br)=C1)C(=O)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)