accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51762-67-5

IUPAC Name: 3-nitrobenzene-1,2-dicarbonitrile
Synonyms: 3-nitrobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 173.13
Molecular Formula: C8H3N3O2
InChi Key: UZJZIZFCQFZDHP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(C#N)=C1C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)