CAS RN 51769-43-8
IUPAC Name:
2-(methylsulfanyl)naphtho[1,2-d][1,3]thiazole
Synonyms:
2-(methylsulfanyl)naphtho[1,2-d][1,3]thiazole
Molecular Weight (g/mol):
231.33
Molecular Formula:
C12H9NS2
InChi Key:
NCJSBVUUUHBCQS-UHFFFAOYSA-N
SMILES:
CSC1=NC2=C(S1)C=CC1=CC=CC=C21
1
–
1
of
1
results
1
2-(Methylthio)naphtho[1,2-d]thiazole 98.0+%, TCI America™
CAS: 51769-43-8 Molecular Formula: C12H9NS2 Molecular Weight (g/mol): 231.331 InChI Key: NCJSBVUUUHBCQS-UHFFFAOYSA-N PubChem CID: 103962 IUPAC Name: 2-methylsulfanylbenzo[e][1,3]benzothiazole SMILES: CSC1=NC2=C(S1)C=CC3=CC=CC=C32