accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5177-27-5

IUPAC Name: 2,4-dichloropyrimidin-5-amine
Synonyms: 2,4-dichloropyrimidin-5-amine
Molecular Weight (g/mol): 163.99
Molecular Formula: C4H3Cl2N3
InChi Key: RINHVELYMZLXIW-UHFFFAOYSA-N
SMILES: NC1=CN=C(Cl)N=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results