accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBr

CAS RN 51776-71-7

IUPAC Name: 1-bromo-4-(difluoromethyl)benzene
Synonyms: 1-bromo-4-(difluoromethyl)benzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: HUSPSWKWFREKSS-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results