accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONHOOCH3CH3H3C

CAS RN 51779-32-9

IUPAC Name: tert-butyl N-[(tert-butoxy)carbonyl]carbamate
Synonyms: tert-butyl N-(tert-butoxycarbonyl)carbamate
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: XCAQIUOFDMREBA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (7)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)