accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOPOOONCH3H3CCH3OOCH3(E/Z)(E/Z)

CAS RN 51779-95-4

IUPAC Name: (2-{[2,3-bis(docos-13-enoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: (2-{[2,3-bis(docos-13-enoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Weight (g/mol): 898.35
Molecular Formula: C52H100NO8P
InChi Key: SDEURMLKLAEUAY-UHFFFAOYNA-N
SMILES: CCCCCCCCC=CCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCC=CCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results