accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 517874-21-4

IUPAC Name: 2-(2,3-dihydro-1,4-benzodioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2,3-dihydro-1,4-benzodioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H19BO4
InChi Key: WIIBAHCYWOUFRM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2OCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results