accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFH3CO

CAS RN 51788-80-8

IUPAC Name: 1-(4-fluoro-2-methoxyphenyl)ethan-1-one
Synonyms: 1-(4-fluoro-2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: YOXBPWVWNQROBJ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results