accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 517918-95-5

IUPAC Name: 1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Synonyms: 1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
Molecular Weight (g/mol): 143.15
Molecular Formula: C8H5N3
InChi Key: DRAQIXNADYAISI-UHFFFAOYSA-N
SMILES: N#CC1=CN=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results