accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NHONH(S)NNH2NHOHN(S)H2NOONH(S)CH3OOHO

CAS RN 51798-45-9

IUPAC Name: (2S)-2-({[(S)-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)({[(1S)-3-carbamoyl-1-{[(2S)-1-oxopropan-2-yl]carbamoyl}propyl]carbamoyl})methyl]carbamoyl}amino)-4-methylpentanoic acid
Synonyms: (2S)-2-({[(S)-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)({[(1S)-3-carbamoyl-1-{[(2S)-1-oxopropan-2-yl]carbamoyl}propyl]carbamoyl})methyl]carbamoyl}amino)-4-methylpentanoic acid
Molecular Weight (g/mol): 512.57
Molecular Formula: C21H36N8O7
InChi Key: IJWCGVPEDDQUDE-WXZHEDMLSA-N
SMILES: CC(C)C[C@H](NC(=O)N[C@@H](C1CCN=C(N)N1)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C)C=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results