accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOHOHO

CAS RN 518-82-1

IUPAC Name: 1,3,8-trihydroxy-6-methyl-9,10-dihydroanthracene-9,10-dione
Synonyms: emodin frangula emodin frangulic acid C.I. natural yellow 14 rheum emodin schuttgelb emodol emodin, rheum persian berry lake emodin, frangula
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: RHMXXJGYXNZAPX-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C=C3C(=O)C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)