accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5182-44-5

IUPAC Name: 2-(3-chlorophenyl)ethan-1-ol
Synonyms: 2-(3-chlorophenyl)ethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: NDWAVJKRSASRPH-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results