CAS RN 51826-90-5
IUPAC Name:
(3-bromopropyl)trimethoxysilane
Synonyms:
(3-bromopropyl)trimethoxysilane
Molecular Weight (g/mol):
243.17
Molecular Formula:
C6H15BrO3Si
InChi Key:
GLISZRPOUBOZDL-UHFFFAOYSA-N
SMILES:
CO[Si](CCCBr)(OC)OC
1
–
1
of
1
results
1
(3-Bromopropyl)trimethoxysilane 98.0+%, TCI America™
CAS: 51826-90-5 Molecular Formula: C6H15BrO3Si Molecular Weight (g/mol): 243.172 MDL Number: MFCD00053894 InChI Key: GLISZRPOUBOZDL-UHFFFAOYSA-N PubChem CID: 103984 IUPAC Name: 3-bromopropyl(trimethoxy)silane SMILES: CO[Si](CCCBr)(OC)OC