accessibility menu, dialog, popup

UserName

Viewing profile as user:
PtOOOOOOH3OH3O

CAS RN 51850-20-5

IUPAC Name: platinum(4+) dihydrogen hexahydroxide
Synonyms: platinum(4+) dihydrogen hexahydroxide
Molecular Weight (g/mol): 299.14
Molecular Formula: H8O6Pt
InChi Key: ILXDKMTYQJUXAB-UHFFFAOYSA-J
SMILES: [H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4]
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)