accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51851-37-7

IUPAC Name: triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Synonyms: perfluorooctyl triethoxysilane
Molecular Weight (g/mol): 510.37
Molecular Formula: C14H19F13O3Si
InChi Key: AVYKQOAMZCAHRG-UHFFFAOYSA-N
SMILES: CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results