accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5187-82-6

BrH3COOSCH3CH3

CAS RN 5187-82-6

IUPAC Name: (2-ethoxy-2-oxoethyl)dimethylsulfanium bromide
Synonyms: (2-ethoxy-2-oxoethyl)dimethylsulfanium bromide
Molecular Weight (g/mol): 229.13
Molecular Formula: C6H13BrO2S
InChi Key: JXFPTJYKYKVENJ-UHFFFAOYSA-M
SMILES: [Br-].CCOC(=O)C[S+](C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)