accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OHOHNONHOOCH3CH3H3C

CAS RN 51871-42-2

IUPAC Name: (2S)-2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)-4-methylpentanoic acid
Synonyms: (2S)-2-{2-[(tert-butoxycarbonyl)amino]acetamido}-4-methylpentanoic acid
Molecular Weight (g/mol): 288.34
Molecular Formula: C13H24N2O5
InChi Key: RGNNFKYULPHSJI-IMWMWJONNA-N
SMILES: CC(C)C[C@H](NC(=O)CNC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)