accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3S

CAS RN 5188-07-8

IUPAC Name: sodium methylsulfanide
Synonyms: sodium methanethiolate
Molecular Weight (g/mol): 70.08
Molecular Formula: CH3NaS
InChi Key: RMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES: [Na+].C[S-]
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
Grade 
  • (4)
  • (4)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (8)
  • (6)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (4)