accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSOHHOOOHO(?)

CAS RN 5189-11-7

IUPAC Name: 1-methyl-4-{6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-2-ylidene}piperidine; 2-hydroxybutanedioic acid
Synonyms: pizotyline malate
Molecular Weight (g/mol): 429.53
Molecular Formula: C23H27NO5S
InChi Key: IWAWCPZVTXCFKD-UHFFFAOYNA-N
SMILES: OC(CC(O)=O)C(O)=O.CN1CCC(CC1)=C1C2=C(CCC3=CC=CC=C13)SC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results