accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiNHNH2OCH3OCH3

CAS RN 51895-58-0

IUPAC Name: (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine
Synonyms: (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine
Molecular Weight (g/mol): 278.47
Molecular Formula: C12H30N2O3Si
InChi Key: AMVXVPUHCLLJRE-UHFFFAOYSA-N
SMILES: CO[Si](CCCNCCCCCCN)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results