accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 519-37-9

IUPAC Name: 7-(2-hydroxyethyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: etofylline ascorphylline bio-phylline etofilina oxphylline (amido) etophyllin oxyphyllin oxytheonyl etophylline oxyethyltheophylline
Molecular Weight (g/mol): 224.22
Molecular Formula: C9H12N4O3
InChi Key: NWPRCRWQMGIBOT-UHFFFAOYSA-N
SMILES: CN1C2=C(N(CCO)C=N2)C(=O)N(C)C1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)